Assay Details
Target Gene Details
Entrez Gene ID: | 15107 |
Gene Name: | hydroxyacyl-Coenzyme A dehydrogenase |
Gene Aliases: |
AA409008, AU019341, AW742602, HCDH, Hadhsc, Schad |
Location: |
Chr.3:131233419-131272101 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hadh | NM_008212.4 | 1 | 104 | NP_032238.2 |
| AK075609.1 | 1 | 31 | ||
| AK132260.1 | 1 | 67 | BAE21064.1 | |
| AK148481.1 | ||||
| AK148486.1 | 1 | 41 | BAE28581.1 | |
| AK167024.1 | 1 | 31 | BAE39197.1 | |
| AK168238.1 | 1 | 51 | BAE40188.1 | |
| AK168877.1 | 1 | 31 | BAE40695.1 | |
| AK169261.1 | 1 | 70 | BAE41023.1 | |
| BC064712.1 | 1 | 46 | AAH64712.1 | |
| D29639.1 | 1 | 30 | BAA06122.1 |

Genomic Map