Assay Details
Target Gene Details
Entrez Gene ID: | 329659 |
Gene Name: | RIKEN cDNA E130311K13 gene |
Gene Aliases: |
- |
Location: |
Chr.3:63914696-63929385 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E130311K13Rik | NM_177856.4 | 1 | 74 | NP_808524.1 |
| AK087526.1 | 1 | 75 | BAC39912.1 | |
| BC125500.1 | 1 | 161 | ||
| BC132160.1 | 1 | 161 |

Genomic Map