Assay Details
Target Gene Details
Entrez Gene ID: | 66614 |
Gene Name: | G patch domain containing 4 |
Gene Aliases: |
2610029K21Rik, Gpatc4 |
Location: |
Chr.3:88043106-88055994 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpatch4 | NM_001110809.2 | NP_001104279.1 | ||
| NM_001286857.1 | 1 | 385 | NP_001273786.1 | |
| NM_025663.4 | NP_079939.3 | |||
| AK011617.1 | ||||
| AK017310.1 | BAB30685.1 | |||
| AK018306.1 | BAB31153.1 | |||
| AK078828.1 | ||||
| AK169111.1 | BAE40893.1 | |||
| BC019490.1 | AAH19490.1 | |||
| BI658293.1 |
More Information
Set Membership: |
|

Genomic Map