Assay Details
Target Gene Details
Entrez Gene ID: | 15168 |
Gene Name: | hyperpolarization-activated, cyclic nucleotide-gated K+ 3 |
Gene Aliases: |
BCNG-4, Bcng4, Hac3, Hcn4 |
Location: |
Chr.3:89146775-89160228 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcn3 | NM_008227.1 | 6 | 1455 | NP_032253.1 |
| AJ225124.1 | 6 | 1455 | CAA12408.1 | |
| AK032225.1 | 6 | 1460 | BAC27769.1 | |
| BC039156.1 | 6 | 1399 | AAH39156.1 | |
| BC138319.1 | 6 | 1429 | ||
| BC138321.1 | 6 | 1429 |
More Information
Set Membership: |
|

Genomic Map