Assay Details
Target Gene Details
Entrez Gene ID: | 15213 |
Gene Name: | hairy/enhancer-of-split related with YRPW motif 1 |
Gene Aliases: |
AI316788, AI414254, CHF2, HRT1, Herp2, Hesr1, bHLHb31, hesr-1 |
Location: |
Chr.3:8663359-8667038 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hey1 | NM_010423.2 | NP_034553.2 | ||
| AB041590.1 | BAA95073.1 | |||
| AF151521.1 | AAD38966.1 | |||
| AF172286.1 | AAF14545.1 | |||
| AF176423.1 | AAD51750.1 | |||
| AF232241.1 | AAF37299.1 | |||
| AJ243895.1 | CAB51321.1 | |||
| AK011149.1 | BAB27432.1 | |||
| AK140212.1 | BAE24283.1 | |||
| BC086635.1 | AAH86635.1 |
More Information
Set Membership: |
|

Genomic Map