Assay Details
Target Gene Details
Entrez Gene ID: | 628324 |
Gene Name: | S100 calcium binding protein A2 |
Gene Aliases: |
CaN19, EG628324, S100L |
Location: |
Chr.3:90590247-90591508 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a2 | NM_001195760.1 | 1 | 50 | NP_001182689.1 |
Target Gene Details
Entrez Gene ID: | 20197 |
Gene Name: | S100 calcium binding protein A3 |
Gene Aliases: |
S100E |
Location: |
Chr.3:90560356-90602706 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a3 | AF087470.1 | AAC64107.1 |
More Information
Set Membership: |
|

Genomic Map