Assay Details
Target Gene Details
Entrez Gene ID: | 51938 |
Gene Name: | coiled-coil domain containing 39 |
Gene Aliases: |
4921507O14Rik, AI447426, AI844750, D3Ertd789e, b2b1304Clo, b2b1735Clo, b2b2025.1Clo |
Location: |
Chr.3:33810757-33844322 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc39 | NM_026222.2 | NP_080498.1 | ||
| AK007153.1 | BAC25166.1 | |||
| AK014839.1 | BAB29574.1 | |||
| AK030087.1 | BAC26777.1 | |||
| BC138917.1 | ||||
| BC138920.1 |
Target Gene Details
Entrez Gene ID: | 67120 |
Gene Name: | tetratricopeptide repeat domain 14 |
Gene Aliases: |
2700016E08Rik, 4930434D01Rik, 4931403I22Rik, 4933402I15Rik, AI662468, AU014779, AW561908, cI-44, mKIAA1980 |
Location: |
Chr.3:33800022-33814860 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc14 | NM_001290500.1 | 11 | 6837 | NP_001277429.1 |
| NM_001290502.1 | 10 | 8873 | NP_001277431.1 | |
| NM_025978.4 | 10 | 8635 | NP_080254.2 | |
| NM_027619.4 | 12 | 6993 | NP_081895.2 | |
| AK040315.1 | 1 | 1722 |

Genomic Map