Assay Details
Target Gene Details
Entrez Gene ID: | 229317 |
Gene Name: | eukaryotic translation initiation factor 2A |
Gene Aliases: |
D030048D22, D3Ertd194e |
Location: |
Chr.3:58525821-58557501 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2a | NM_001005509.2 | NP_001005509.1 | ||
| AK050971.1 | ||||
| AK078519.1 | BAC37320.1 | |||
| AK145275.1 | ||||
| BC030447.1 | ||||
| BC082557.1 | AAH82557.1 |
More Information
Set Membership: |
|

Genomic Map