Assay Details
Target Gene Details
Entrez Gene ID: | 320478 |
Gene Name: | SOX2 overlapping transcript (non-protein coding) |
Gene Aliases: |
B230215L15Rik |
Location: |
Chr.3:34560381-34677993 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox2ot | NR_015580.2 |
More Information
Set Membership: |
|

Genomic Map