Assay Details
Target Gene Details
Entrez Gene ID: | 78523 |
Gene Name: | mitochondrial ribosomal protein L9 |
Gene Aliases: |
8030480E20Rik, AA409733, C330013D18Rik, L9mt |
Location: |
Chr.3:94443272-94448708 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl9 | NM_030116.2 | NP_084392.1 | ||
| AB049637.1 | BAB40842.1 | |||
| AK021196.1 | ||||
| AK040055.1 | BAC30503.1 | |||
| AK046099.1 | BAC32604.1 | |||
| AK161563.1 | ||||
| AK162023.1 | BAE36685.1 | |||
| BC116385.1 | ||||
| BC116386.1 |
More Information
Set Membership: |
|

Genomic Map