Assay Details
Target Gene Details
Entrez Gene ID: | 192167 |
Gene Name: | neuroligin 1 |
Gene Aliases: |
6330415N05Rik, BB179718, NL1, Nlg1, mKIAA1070 |
Location: |
Chr.3:25431841-26332056 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlgn1 | NM_138666.3 | NP_619607.2 | ||
| AK083116.1 | ||||
| AK122433.1 | BAC65715.1 |
More Information
Set Membership: |
|

Genomic Map