Assay Details
Target Gene Details
Entrez Gene ID: | 269437 |
Gene Name: | phospholipase C, eta 1 |
Gene Aliases: |
BC042549, PLCeta1, Plcl3 |
Location: |
Chr.3:63696232-63899677 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plch1 | NM_001177732.1 | 7 | 969 | NP_001171203.1 |
| NM_001177733.1 | 7 | 969 | NP_001171204.1 | |
| NM_183191.3 | 7 | 969 | NP_899014.2 | |
| AY691172.1 | 7 | 969 | AAW22609.1 | |
| AY691173.1 | 7 | 969 | AAW22610.1 | |
| AY691174.1 | 7 | 969 | AAW22611.1 | |
| BC042549.1 | AAH42549.1 | |||
| BC052372.1 | AAH52372.1 | |||
| BC055005.1 | 7 | 1046 | AAH55005.1 |
More Information
Set Membership: |
|

Genomic Map