Assay Details
Target Gene Details
Entrez Gene ID: | 20196 |
Gene Name: | S100 calcium binding protein A13 |
Gene Aliases: |
- |
Location: |
Chr.3:90514435-90524581 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a13 | NM_009113.4 | 4 | 629 | NP_033139.2 |
| AK002755.1 | 3 | 428 | BAB22332.1 | |
| AK008895.1 | 3 | 428 | BAB25958.1 | |
| BC005687.1 | 4 | 629 | AAH05687.1 | |
| CK334302.1 | ||||
| CX219167.1 | 3 | 352 | ||
| D18141.1 | ||||
| X99921.1 | 3 | 356 | CAA68189.1 |
More Information
Set Membership: |
|

Genomic Map