Assay Details
Target Gene Details
Entrez Gene ID: | 20713 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade I, member 1 |
Gene Aliases: |
AI837402, Ns, PI-12, PI12, Spi17 |
Location: |
Chr.3:75557503-75642523 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpini1 | NM_009250.2 | NP_033276.1 | ||
| AJ001700.1 | CAA04939.1 | |||
| AK032152.1 | BAC27727.1 | |||
| AK133770.1 | ||||
| AK164355.1 | 2 | 1476 | ||
| BC006776.1 | AAH06776.1 |
More Information
Set Membership: |
|

Genomic Map