Assay Details
Target Gene Details
Entrez Gene ID: | 67931 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade I, member 2 |
Gene Aliases: |
Spi14 |
Location: |
Chr.3:75242357-75270078 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpini2 | NM_026460.3 | 3 | 382 | NP_080736.2 |
| AF251276.1 | 3 | 367 | AAF65821.1 | |
| AK007347.1 | 3 | 383 | BAB24976.1 | |
| AK007510.1 | 3 | 357 | BAB25079.1 | |
| AK008931.1 | 3 | 383 | ||
| BC098466.1 | 3 | 359 | AAH98466.1 |
More Information
Set Membership: |
|

Genomic Map