Assay Details
Target Gene Details
Entrez Gene ID: | 20197 |
Gene Name: | S100 calcium binding protein A3 |
Gene Aliases: |
S100E |
Location: |
Chr.3:90560356-90602706 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a3 | NM_011310.2 | 2 | 122 | NP_035440.1 |
| AF004941.1 | 2 | 122 | AAC78730.1 | |
| AF087470.1 | 2 | 97 | AAC64107.1 | |
| AK133717.1 | 2 | 124 | BAE21797.1 | |
| AK161105.1 | 1 | 1604 | ||
| BC099893.1 | 2 | 119 | AAH99893.1 | |
| BC100690.1 | 2 | 119 | AAI00691.1 | |
| BC100696.1 | 2 | 119 | AAI00697.1 | |
| BC100697.1 | 2 | 119 | AAI00698.1 |
More Information
Set Membership: |
|

Genomic Map