Assay Details
Target Gene Details
Entrez Gene ID: | 20198 |
Gene Name: | S100 calcium binding protein A4 |
Gene Aliases: |
18A2, 42a, Capl, FSp1, Mts1, PeL98, metastasin, pk9a |
Location: |
Chr.3:90603770-90606045 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a4 | NM_011311.2 | 2 | 73 | NP_035441.1 |
| AK003065.1 | 2 | 79 | BAB22543.1 | |
| AK160522.1 | 2 | 74 | BAE35841.1 | |
| BC051214.1 | AAH51214.1 | |||
| BC094635.1 | AAH94635.1 | |||
| BX526220.1 | ||||
| D00208.1 | 2 | 73 | BAA00148.1 | |
| X05835.1 | CAA29282.1 | |||
| X16190.1 | 1 | 32 | CAA34316.1 |
More Information
Set Membership: |
|

Genomic Map