Assay Details
Target Gene Details
Entrez Gene ID: | 381493 |
Gene Name: | S100 calcium binding protein A7A |
Gene Aliases: |
AY465109, Gm1020, S100A7L1, S100A7f, S100a15, S100a15a, S100a17l1 |
Location: |
Chr.3:90651934-90658130 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a7a | NM_199422.1 | NP_955454.1 | ||
| AY465109.1 | AAR32782.1 | |||
| AY582964.1 | AAS91715.1 | |||
| BC107307.1 | AAI07308.1 | |||
| BC107308.1 | AAI07309.1 |
More Information
Set Membership: |
|

Genomic Map