Assay Details
Target Gene Details
Entrez Gene ID: | 108943 |
Gene Name: | tRNA methyltransferase 10A |
Gene Aliases: |
3110023L08Rik, AA794508, Rg9mtd2, Rnmtd2 |
Location: |
Chr.3:138143517-138159821 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt10a | NM_175389.4 | NP_780598.2 | ||
| AK089967.1 | BAC41020.1 | |||
| AK145681.1 | ||||
| AK146481.1 | ||||
| BC022121.1 | AAH22121.1 | |||
| BC090650.1 | AAH90650.1 | |||
| BC098478.1 | AAH98478.1 | |||
| BC141035.1 |
More Information
Set Membership: |
|

Genomic Map