Assay Details
Target Gene Details
Entrez Gene ID: | 72634 |
Gene Name: | tudor and KH domain containing protein |
Gene Aliases: |
2700091C21Rik, Tdrd2 |
Location: |
Chr.3:94413239-94432975 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdrkh | NM_028307.1 | NP_082583.1 | ||
| AK012596.1 | ||||
| BC049363.1 | AAH49363.1 | |||
| BC057030.1 | AAH57030.1 |
More Information
Set Membership: |
|

Genomic Map