Assay Details
Target Gene Details
Entrez Gene ID: | 18857 |
Gene Name: | peripheral myelin protein 2 |
Gene Aliases: |
P2 |
Location: |
Chr.3:10179851-10183885 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmp2 | NM_001030305.2 | 1 | 38 | NP_001025476.1 |
| BC099520.1 | 1 | 38 | AAH99520.1 |

Genomic Map