Assay Details
Target Gene Details
Entrez Gene ID: | 27059 |
Gene Name: | SH3 domain protein D19 |
Gene Aliases: |
AW011754, Kryn |
Location: |
Chr.3:85970956-86130522 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3d19 | NM_001082414.2 | NP_001075883.2 | ||
| AF003234.1 | AAB82091.1 | |||
| AK050185.1 | BAC34114.1 | |||
| BC031117.1 | AAH31117.1 | |||
| D89677.2 | BAA19686.2 |

Genomic Map