Assay Details
Target Gene Details
Entrez Gene ID: | 75778 |
Gene Name: | thioesterase superfamily member 4 |
Gene Aliases: |
2700077M13Rik, 4921507I02Rik |
Location: |
Chr.3:94310106-94332540 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Them4 | NM_029431.1 | NP_083707.1 | ||
| AK014834.1 | 4 | 527 | BAB29572.1 | |
| AK038420.1 | 4 | 492 | BAC29995.1 | |
| AK138397.1 | BAE23642.1 | |||
| BC022612.1 | AAH22612.1 | |||
| BC134396.1 | 4 | 590 |
More Information
Set Membership: |
|

Genomic Map