Assay Details
Target Gene Details
Entrez Gene ID: | 94212 |
Gene Name: | phosphoprotein associated with glycosphingolipid microdomains 1 |
Gene Aliases: |
Cbp, F730007C19Rik, Pag |
Location: |
Chr.3:9687479-9833679 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pag1 | NM_001195031.1 | 6 | 906 | NP_001181960.1 |
| NM_053182.5 | 5 | 847 | NP_444412.2 | |
| AF250192.1 | 3 | 228 | AAG44565.1 | |
| AK040230.1 | 6 | 856 | BAC30545.1 | |
| AK076515.1 | 2 | 345 | ||
| AK089329.1 | 5 | 799 | BAC40842.1 | |
| AK156000.1 | 6 | 858 | BAE33539.1 | |
| AK156986.1 | 6 | 831 | ||
| AK172106.1 | 6 | 829 | BAE42829.1 | |
| BC145328.1 | 6 | 730 | ||
| BC145761.1 | 6 | 730 | ||
| BC145763.1 | 6 | 730 |

Genomic Map