Assay Details
Target Gene Details
Entrez Gene ID: | 20416 |
Gene Name: | src homology 2 domain-containing transforming protein C1 |
Gene Aliases: |
Shc, ShcA, p66, p66shc |
Location: |
Chr.3:89418481-89430029 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shc1 | NM_011368.5 | NP_035498.2 | ||
| AK049357.1 | BAC33706.1 | |||
| BC036172.1 | AAH36172.1 | |||
| BY341731.1 | ||||
| U15784.1 | AAC52146.1 |
More Information
Set Membership: |
|

Genomic Map