Assay Details
Target Gene Details
Entrez Gene ID: | 56426 |
Gene Name: | programmed cell death 10 |
Gene Aliases: |
2410003B13Rik, Ccm3, Tfa15, Tfar15 |
Location: |
Chr.3:75516490-75556852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdcd10 | NM_019745.4 | NP_062719.2 | ||
| AF159368.1 | AAD44335.1 | |||
| AK005594.1 | BAB24143.1 | |||
| AK075926.1 | BAC36058.1 | |||
| AK077308.1 | BAC36742.1 | |||
| AK137198.1 | BAE23263.1 | |||
| AK149182.1 | ||||
| AK160234.1 | BAE35705.1 | |||
| AK168857.1 | BAE40678.1 | |||
| AK169689.1 | BAE41306.1 | |||
| BC019650.1 | AAH19650.1 | |||
| BC086778.1 | AAH86778.1 |

Genomic Map