Assay Details
Target Gene Details
Entrez Gene ID: | 211666 |
Gene Name: | microsomal glutathione S-transferase 2 |
Gene Aliases: |
GST2, MGST-II |
Location: |
Chr.3:51660360-51682675 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgst2 | NM_174995.2 | NP_778160.2 | ||
| AA919832.1 | ||||
| BC132234.1 | ||||
| BC132260.1 | ||||
| BX639527.1 |
More Information
Set Membership: |
|

Genomic Map