Assay Details
Target Gene Details
Entrez Gene ID: | 229228 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 6 |
Gene Aliases: |
Asfgf2b |
Location: |
Chr.3:37404982-37420186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt6 | NM_001291044.1 | NP_001277973.1 | ||
| NM_153561.3 | NP_705789.1 | |||
| AF453427.1 | AAN76847.1 | |||
| AF453428.1 | AAN76848.1 | |||
| AK145299.1 | BAE26354.1 | |||
| BC027267.1 | AAH27267.1 | |||
| BC146000.1 |
More Information
Set Membership: |
|

Genomic Map