Assay Details
Target Gene Details
Entrez Gene ID: | 18741 |
Gene Name: | paired-like homeodomain transcription factor 2 |
Gene Aliases: |
9430085M16Rik, Brx1, Brx1b, Munc30, Otlx2, Ptx2, Rieg |
Location: |
Chr.3:129199878-129219594 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitx2 | NM_001042504.2 | 1 | 160 | NP_001035969.1 |
| NM_001286942.1 | 1 | 160 | NP_001273871.1 | |
| NM_011098.4 | 1 | 160 | NP_035228.2 | |
| AB006321.1 | 1 | 120 | BAA75248.1 | |
| AF048723.1 | 1 | 97 | AAC40086.1 | |
| BY775551.1 | 1 | 160 | ||
| U80010.1 | 1 | 95 | AAC53119.1 | |
| U80011.1 | 1 | 113 | AAC53120.1 | |
| U80036.1 | 1 | 131 | AAB38505.1 |
More Information
Set Membership: |
|

Genomic Map