Assay Details
Target Gene Details
Entrez Gene ID: | 18767 |
Gene Name: | protein kinase inhibitor, alpha |
Gene Aliases: |
AI415001, PKIalpha |
Location: |
Chr.3:7366604-7445366 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkia | NM_008862.3 | NP_032888.1 | ||
| AK139347.1 | BAE23970.1 | |||
| BC048244.1 | AAH48244.1 | |||
| BY253073.1 |
More Information
Set Membership: |
|

Genomic Map