Assay Details
Target Gene Details
Entrez Gene ID: | 18767 |
Gene Name: | protein kinase inhibitor, alpha |
Gene Aliases: |
AI415001, PKIalpha |
Location: |
Chr.3:7366604-7445366 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkia | NM_008862.3 | 2 | 278 | NP_032888.1 |
| AK139347.1 | 2 | 210 | BAE23970.1 | |
| AK190537.1 | 1 | 83 | ||
| AK190735.1 | 1 | 83 | ||
| AK193224.1 | 1 | 83 | ||
| BC048244.1 | 2 | 212 | AAH48244.1 | |
| BY253073.1 | 2 | 279 | ||
| M63554.1 | 1 | 131 | AAA39940.1 |
More Information
Set Membership: |
|

Genomic Map