Assay Details
Target Gene Details
Entrez Gene ID: | 665113 |
Gene Name: | TRAF2 and NCK interacting kinase |
Gene Aliases: |
1500031A17Rik, 4831440I19Rik, AI451411, C530008O15Rik, C630040K21Rik |
Location: |
Chr.3:28263214-28670585 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnik | NM_001163007.1 | 18 | 2499 | NP_001156479.1 |
| NM_001163008.1 | 17 | 2412 | NP_001156480.1 | |
| NM_001163009.1 | 18 | 2499 | NP_001156481.1 | |
| NM_026910.1 | 18 | 2499 | NP_081186.1 | |
| BC137799.1 | 18 | 2150 | ||
| BC137800.1 | 18 | 2150 | ||
| BC151003.1 | 18 | 2150 | ||
| BC158011.1 | 17 | 2063 | ||
| BC158059.1 | 18 | 2150 | ||
| CD348964.1 | 2 | 74 |
More Information
Set Membership: |
|

Genomic Map