Assay Details
Target Gene Details
Entrez Gene ID: | 230163 |
Gene Name: | aldolase B, fructose-bisphosphate |
Gene Aliases: |
Aldo-2, Aldo2, BC016435 |
Location: |
Chr.4:49535995-49549483 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldob | NM_144903.2 | 7 | 818 | NP_659152.1 |
| AK135385.1 | 7 | 847 | BAE22514.1 | |
| AK149395.1 | 7 | 835 | BAE28850.1 | |
| AK149598.1 | 7 | 832 | BAE28983.1 | |
| AK167334.1 | 7 | 835 | BAE39436.1 | |
| AK167566.1 | 7 | 832 | BAE39629.1 | |
| AK168600.1 | 7 | 832 | BAE40467.1 | |
| AK168997.1 | 7 | 882 | BAE40796.1 | |
| BC016435.1 | 7 | 814 | AAH16435.1 | |
| BC022113.1 | 7 | 791 | AAH22113.1 | |
| BC024056.1 | 7 | 824 | AAH24056.1 | |
| BC024112.1 | 7 | 823 | AAH24112.1 | |
| BC026577.1 | 7 | 783 | AAH26577.1 | |
| BC030724.1 | 7 | 820 | AAH30724.1 | |
| BC030725.1 | 7 | 824 | AAH30725.1 | |
| BC034169.1 | 7 | 824 | AAH34169.1 | |
| BC034171.1 | 7 | 801 | AAH34171.1 | |
| BC034172.1 | 7 | 788 | AAH34172.1 | |
| BC034173.1 | 7 | 800 | AAH34173.1 | |
| BC036130.1 | 7 | 801 | AAH36130.1 | |
| BC036131.1 | 7 | 818 | AAH36131.1 | |
| BC036132.1 | 7 | 818 | AAH36132.1 | |
| BC036133.1 | 7 | 830 | AAH36133.1 |
More Information
Set Membership: |
|

Genomic Map