Assay Details
Target Gene Details
Entrez Gene ID: | 12319 |
Gene Name: | carbonic anhydrase 8 |
Gene Aliases: |
AW546993, Ca8, Cals, Cals1, Carp, wdl |
Location: |
Chr.4:8141493-8239041 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car8 | NM_007592.3 | NP_031618.2 | ||
| AK004896.1 | BAC25100.1 | |||
| AK004923.1 | ||||
| BC010773.1 | AAH10773.1 | |||
| EU234006.1 |

Genomic Map