Assay Details
Target Gene Details
Entrez Gene ID: | 329942 |
Gene Name: | CUB and Sushi multiple domains 2 |
Gene Aliases: |
B230311I21, B230378I17, Gm139, mKIAA1884 |
Location: |
Chr.4:127988016-128567666 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csmd2 | NM_001281955.1 | NP_001268884.1 | ||
| AK046385.1 | ||||
| AK140129.1 |
Target Gene Details
Entrez Gene ID: | 74989 |
Gene Name: | CUB and Sushi multiple domains 2, opposite strand |
Gene Aliases: |
4930465A12Rik |
Location: |
Chr.4:128132201-128166597 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csmd2os | NM_029137.1 | 5 | 528 | NP_083413.1 |
| AK015502.1 | 5 | 529 |
More Information
Set Membership: |
|

Genomic Map