Assay Details
Target Gene Details
Entrez Gene ID: | 11303 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 1 |
Gene Aliases: |
ABC-1, Abc1 |
Location: |
Chr.4:53030789-53159988 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca1 | NM_013454.3 | 27 | 4169 | NP_038482.3 |
| AK051920.1 | 27 | 4170 | BAC34811.1 | |
| X75926.1 | 26 | 3932 | CAA53530.1 |
More Information
Set Membership: |
|

Genomic Map