Assay Details
Target Gene Details
Entrez Gene ID: | 11699 |
Gene Name: | alpha 1 microglobulin/bikunin |
Gene Aliases: |
AI194774, ASPI, HI-30, Intin4, Itil, UTI |
Location: |
Chr.4:63143279-63154142 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ambp | NM_007443.3 | NP_031469.1 | ||
| AK004907.1 | BAB23659.1 | |||
| BC021660.1 | AAH21660.1 | |||
| D28812.1 | BAA05973.1 | |||
| X68680.1 | CAA48640.1 |

Genomic Map