Assay Details
Target Gene Details
Entrez Gene ID: | 100317 |
Gene Name: | expressed sequence AU040320 |
Gene Aliases: |
A730047D20Rik, Kiaa0319l |
Location: |
Chr.4:126753555-126869694 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU040320 | NM_001035525.1 | NP_001030602.1 | ||
| AK049570.1 | BAC33818.1 | |||
| AK084668.1 | BAC39244.1 | |||
| AK135027.1 | 1 | 417 | ||
| AK172674.1 | 1 | 106 |

Genomic Map