Assay Details
Target Gene Details
Entrez Gene ID: | 72607 |
Gene Name: | ubiquitin specific peptidase 13 (isopeptidase T-3) |
Gene Aliases: |
2700071E21Rik, AI848077, ISOT3, IsoT-3 |
Location: |
Chr.3:32817019-32938075 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp13 | NM_001013024.2 | NP_001013042.1 | ||
| AK135141.1 | ||||
| BC090999.1 | AAH90999.1 |
More Information
Set Membership: |
|

Genomic Map