Assay Details
Target Gene Details
Entrez Gene ID: | 230379 |
Gene Name: | alkaline ceramidase 2 |
Gene Aliases: |
2410116I05Rik, Asah3l, CRG-L1, maCER2 |
Location: |
Chr.4:86874342-86932490 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acer2 | NM_001290541.1 | 1 | 118 | NP_001277470.1 |
| NM_001290543.1 | 1 | 118 | NP_001277472.1 | |
| NM_139306.3 | 1 | 118 | NP_647467.1 | |
| AF282864.1 | AAL40408.1 | |||
| AK085306.1 | 1 | 114 | BAC39416.1 | |
| AY312515.1 | AAQ85131.1 | |||
| BC059819.1 | 1 | 118 | AAH59819.1 | |
| CB723138.1 | 1 | 118 |

Genomic Map