Assay Details
Target Gene Details
Entrez Gene ID: | 66209 |
Gene Name: | INTS3 and NABP interacting protein |
Gene Aliases: |
1110054O05Rik, 2610312O17Rik, AA399876, Ssbip1 |
Location: |
Chr.4:59769642-59783855 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inip | NM_001013577.1 | NP_001013595.1 | ||
| AK004256.1 | BAC25075.1 | |||
| AK005245.1 | ||||
| AK012023.1 | ||||
| AK049428.1 | ||||
| AK159116.1 | BAE34832.1 | |||
| AK169537.1 | BAE41217.1 | |||
| BB641174.1 | ||||
| BC116204.1 | ||||
| BC116205.1 |

Genomic Map