Assay Details
Target Gene Details
Entrez Gene ID: | 67120 |
Gene Name: | tetratricopeptide repeat domain 14 |
Gene Aliases: |
2700016E08Rik, 4930434D01Rik, 4931403I22Rik, 4933402I15Rik, AI662468, AU014779, AW561908, cI-44, mKIAA1980 |
Location: |
Chr.3:33800022-33814860 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc14 | NM_001290500.1 | NP_001277429.1 | ||
| NM_001290502.1 | NP_001277431.1 | |||
| NM_025978.4 | NP_080254.2 | |||
| NM_027619.4 | NP_081895.2 | |||
| AK012246.1 | BAB28115.1 | |||
| AK015308.1 | ||||
| AK016619.1 | ||||
| AK017763.1 | BAB30914.1 | |||
| AK031605.1 | BAC27473.1 | |||
| AK035570.1 | BAC29109.1 | |||
| AK041095.1 | BAC30819.1 | |||
| AK044266.1 | ||||
| AK134539.1 | ||||
| AK173315.1 | BAD32593.1 | |||
| BC021448.1 | ||||
| BC024847.1 | AAH24847.1 | |||
| BC138212.1 | ||||
| BC145944.1 |

Genomic Map