Assay Details
Target Gene Details
Entrez Gene ID: | 97064 |
Gene Name: | WW domain containing transcription regulator 1 |
Gene Aliases: |
2310058J06Rik, 2610021I22Rik, C78399, Taz |
Location: |
Chr.3:57455644-57575910 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wwtr1 | NM_001168281.1 | 3 | 709 | NP_001161753.1 |
| NM_133784.3 | 3 | 788 | NP_598545.2 | |
| AJ299430.1 | 3 | 723 | CAC17733.1 | |
| AK009983.1 | 3 | 721 | ||
| AK011504.1 | 3 | 764 | ||
| AK142299.1 | 3 | 710 | BAE25019.1 | |
| AK142720.1 | 3 | 713 | BAE25174.1 | |
| BC004640.1 | 3 | 709 | AAH04640.1 | |
| BC014727.1 | 3 | 740 | AAH14727.1 |
More Information
Set Membership: |
|

Genomic Map