Assay Details
Target Gene Details
Entrez Gene ID: | 56079 |
Gene Name: | astrotactin 2 |
Gene Aliases: |
1d8, Astnl, bM452J22.1 |
Location: |
Chr.4:65380803-66404530 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Astn2 | NM_019514.3 | 11 | 2134 | NP_062387.2 |
| NM_207109.2 | 12 | 2290 | NP_996992.1 | |
| AF116573.1 | 9 | 1302 | AAF14356.1 | |
| AK041160.1 | 13 | 2194 | BAC30843.1 |

Genomic Map