Assay Details
Target Gene Details
Entrez Gene ID: | 214191 |
Gene Name: | tetratricopeptide repeat domain 24 |
Gene Aliases: |
A430025D11Rik |
Location: |
Chr.3:88069410-88081987 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc24 | NM_172526.3 | 8 | 1412 | NP_766114.1 |
| AK039884.1 | 8 | 1413 | BAC30472.1 | |
| AK076743.1 | 3 | 228 | ||
| BC055114.1 | 3 | 227 | AAH55114.1 | |
| BC147554.1 | 7 | 1116 | ||
| BC147555.1 | 7 | 1116 |

Genomic Map