Assay Details
Target Gene Details
Entrez Gene ID: | 66469 |
Gene Name: | family with sequence similarity 213, member B |
Gene Aliases: |
2810405K02Rik, AI836168, PM/PGFS |
Location: |
Chr.4:154896430-154899043 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam213b | NM_025582.3 | NP_079858.2 | ||
| AK005188.1 | BAB23870.1 | |||
| AK012999.1 | ||||
| BC030453.1 | AAH30453.1 |
More Information
Set Membership: |
|

Genomic Map