Assay Details
Target Gene Details
Entrez Gene ID: | 230235 |
Gene Name: | ferric-chelate reductase 1 like |
Gene Aliases: |
6430704M03Rik |
Location: |
Chr.4:56960136-56990391 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frrs1l | NM_001142965.1 | 4 | 633 | NP_001136437.1 |
| AK038461.1 | 4 | 337 | BAC30008.1 | |
| AK078331.1 | 3 | 296 | BAC37221.1 | |
| BC132345.1 | 4 | 660 |

Genomic Map