Assay Details
Target Gene Details
Entrez Gene ID: | 108888 |
Gene Name: | ATPase family, AAA domain containing 3A |
Gene Aliases: |
2400004H09Rik, Atad3, Tob3, mKIAA1273 |
Location: |
Chr.4:155740640-155761108 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atad3a | NM_179203.3 | NP_849534.2 | ||
| AF343079.1 | AAK38648.1 | |||
| AK075921.1 | BAC36055.1 | |||
| AK148974.1 | BAE28707.1 | |||
| AK149702.1 | BAE29037.1 | |||
| AK150037.1 | BAE29259.1 | |||
| AK165724.1 | ||||
| AK166099.1 | BAE38570.1 | |||
| AK172038.1 | BAE42791.1 | |||
| AK172350.1 | ||||
| AK173141.1 | BAD32419.1 | |||
| AK181017.1 | ||||
| BC023301.1 | AAH23301.1 | |||
| BC058373.1 | AAH58373.1 | |||
| BC060036.1 | AAH60036.1 |

Genomic Map