Assay Details
Target Gene Details
Entrez Gene ID: | 66259 |
Gene Name: | calcium/calmodulin-dependent protein kinase II inhibitor 1 |
Gene Aliases: |
1810006K23Rik, AI848599, AI891706 |
Location: |
Chr.4:138455148-138460126 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2n1 | NM_025451.2 | 1 | 108 | NP_079727.1 |
| AY523601.1 | 1 | 70 | AAS02090.1 |
More Information
Set Membership: |
|

Genomic Map