Assay Details
Target Gene Details
Entrez Gene ID: | 230234 |
Gene Name: | family with sequence similarity 206, member A |
Gene Aliases: |
simiate |
Location: |
Chr.4:56802329-56809605 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam206a | NM_001081420.1 | NP_001074889.1 | ||
| AK043588.1 | 1 | 1233 | ||
| AK052647.1 | BAC35079.1 | |||
| AK164596.1 | 2 | 193 | ||
| BC026590.1 | ||||
| BC048548.1 |

Genomic Map